CAS RN 215434-39-2
CAS RN 215434-39-2
IUPAC Name:
2-{4-[5-chloro-3-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}ethan-1-ol
Synonyms:
2-{4-[5-chloro-3-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}ethanol
Molecular Weight (g/mol):
309.72
Molecular Formula:
C12H15ClF3N3O
InChi Key:
NYMGNJNWMTZCFC-UHFFFAOYSA-N
SMILES:
OCCN1CCN(CC1)C1=NC=C(Cl)C=C1C(F)(F)F