accessibility menu, dialog, popup

UserName

HOClN

CAS RN 21543-49-7

IUPAC Name: (6-chloropyridin-3-yl)methanol
Synonyms: (6-chloropyridin-3-yl)methanol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: GOXYBEXWMJZLJB-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Cl)N=C1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results