CAS RN 21430-85-3
CAS RN 21430-85-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichlorobis(ethylenediamine)platinum(II), Pt 50%
CAS: 21430-85-3 Molecular Formula: C4H16Cl2N4Pt Molecular Weight (g/mol): 386.18 MDL Number: MFCD00009881 InChI Key: MXLIXEZAMQLDMM-UHFFFAOYSA-L Synonym: Bis(ethylenediamine)platinum(II) chloride IUPAC Name: platinum(2+) bis(ethane-1,2-diamine) dichloride SMILES: [Cl-].[Cl-].[Pt++].NCCN.NCCN