CAS RN 213690-27-8
CAS RN 213690-27-8
IUPAC Name:
4-bromo-6-(2,5-dihydro-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Synonyms:
4-bromo-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol):
240.06
Molecular Formula:
C9H6BrNO2
InChi Key:
GBXOJTWVFYSJRI-UHFFFAOYSA-N
SMILES:
BrC1=CC(=C2ONC=C2)C(=O)C=C1
1
–
1
of
1
results
1
4-Bromo-2-(5-isoxazolyl)phenol, 97%
CAS: 213690-27-8 Molecular Formula: C9H6BrNO2 Molecular Weight (g/mol): 240.06 MDL Number: MFCD00799493 InChI Key: GBXOJTWVFYSJRI-UHFFFAOYSA-N Synonym: 4-bromo-2-5-isoxazolyl phenol,4-bromo-6-2h-1,2-oxazol-5-ylidene cyclohexa-2,4-dien-1-one,4-bromo-2-1,2-oxazol-5-yl phenol,acmc-1ckhc,4-bromo-2-isoxazole-5-yl phenol,phenol,4-bromo-2-5-isoxazolyl PubChem CID: 6744794 IUPAC Name: 4-bromo-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one SMILES: BrC1=CC(=C2ONC=C2)C(=O)C=C1