CAS RN 213270-36-1
IUPAC Name:
(3S)-3-(methoxycarbonyl)-5-methylhexanoate
Synonyms:
(3S)-3-(methoxycarbonyl)-5-methylhexanoate
Molecular Weight (g/mol):
187.22
Molecular Formula:
C9H15O4
InChi Key:
AJNNFMRCMKGYHA-ZETCQYMHSA-M
SMILES:
COC(=O)[C@@H](CC(C)C)CC([O-])=O
1
–
1
of
1
results
1
(S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific™
CAS: 213270-36-1 Molecular Formula: C9H16O4 Molecular Weight (g/mol): 188.22 MDL Number: MFCD01091033 InChI Key: AJNNFMRCMKGYHA-ZETCQYMHSA-N Synonym: s-3-methoxycarbonyl-5-methylhexanoic acid,s---2-isobutylsuccinic acid 1-methyl ester,3s-3-methoxycarbonyl-5-methylhexanoic acid,s-2-isobutylsuccinic acid 1-methyl ester,s-2-2-methylpropyl butanedioic acid 1-methyl ester,butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s PubChem CID: 2734513 IUPAC Name: (3S)-3-methoxycarbonyl-5-methylhexanoic acid SMILES: CC(C)CC(CC(=O)O)C(=O)OC