CAS RN 211947-56-7
CAS RN 211947-56-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Nonyl Phenol-13C6, TRC
CAS: 211947-56-7 Molecular Formula: 13C6 C9 H24 O Molecular Weight (g/mol): 226.31 Synonym: p-Nonylhenol-13C6,4-Nonylphenol-13C6,4-n-Nonyl Phenol-13C6,p-NP-13C6,p-Nonylphenol-13C6,p-n-Nonylphenol-13C6 IUPAC Name: 4-nonyl(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-ol SMILES: CCCCCCCCC[13c]1[13cH][13cH][13c](O)[13cH][13cH]1