CAS RN 21182-09-2
CAS RN 21182-09-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,3'-(Pyridin-4-ylmethylene)bis(1H-indole), TRC
CAS: 21182-09-2 Molecular Formula: C22H17N3 Molecular Weight (g/mol): 323.39 Synonym: 3,3'-(4-Pyridinylmethylene)di(indole) IUPAC Name: 3-[1H-indol-3-yl(pyridin-4-yl)methyl]-1H-indole SMILES: c1ccc2c(c[nH]c2c1)C(c3ccncc3)c4c[nH]c5ccccc45