accessibility menu, dialog, popup

UserName

CAS RN 21169-71-1

IUPAC Name: 1,2-oxazole-5-carboxylate
Synonyms: 1,2-oxazole-5-carboxylate
Molecular Weight (g/mol): 112.07
Molecular Formula: C4H2NO3
InChi Key: MIIQJAUWHSUTIT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=NO1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results