accessibility menu, dialog, popup

UserName

CAS RN 2109-66-2

CH3HONO

CAS RN 2109-66-2

IUPAC Name: 1-(morpholin-4-yl)propan-2-ol
Synonyms: 1-(morpholin-4-yl)propan-2-ol
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: YAXQOLYGKLGQKA-UHFFFAOYNA-N
SMILES: CC(O)CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results