accessibility menu, dialog, popup

UserName

CAS RN 209538-87-4

IUPAC Name: ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-(3-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 263.31
Molecular Formula: C13H13NO3S
InChi Key: RBLKTMVSJHHLGF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1=CC(O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results