CAS RN 209481-20-9
CAS RN 209481-20-9
IUPAC Name:
5-chloro-N-[4-methoxy-3-(piperazin-1-yl)phenyl]-3-methyl-1-benzothiophene-2-sulfonamide hydrochloride
Synonyms:
5-chloro-N-[4-methoxy-3-(piperazin-1-yl)phenyl]-3-methyl-1-benzothiophene-2-sulfonamide hydrochloride
Molecular Weight (g/mol):
488.44
Molecular Formula:
C20H23Cl2N3O3S2
InChi Key:
RMXZRJYGJMSDQK-UHFFFAOYSA-N
SMILES:
Cl.COC1=C(C=C(NS(=O)(=O)C2=C(C)C3=CC(Cl)=CC=C3S2)C=C1)N1CCNCC1
1
–
1
of
1
results
1
Tocris Bioscience™ SB 271046 hydrochloride
CAS: 209481-20-9 Molecular Formula: C20H23Cl2N3O3S2