accessibility menu, dialog, popup

UserName

H3COHNO

CAS RN 20928-63-6

IUPAC Name: 3-methoxy-1,2-dihydropyridin-2-one
Synonyms: 3-methoxy-1H-pyridin-2-one
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: LKIMDXQLHFCXQF-UHFFFAOYSA-N
SMILES: COC1=CC=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)