accessibility menu, dialog, popup

UserName

CAS RN 20845-34-5

CH3NHO

CAS RN 20845-34-5

IUPAC Name: (1-methylpiperidin-2-yl)methanol
Synonyms: (1-methylpiperidin-2-yl)methanol
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: HXXJMMLIEYAFOZ-UHFFFAOYNA-N
SMILES: CN1CCCCC1CO
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)