accessibility menu, dialog, popup

UserName

CAS RN 208108-76-3

SOHOFFF

CAS RN 208108-76-3

IUPAC Name: 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Synonyms: 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 272.24
Molecular Formula: C12H7F3O2S
InChi Key: GIUAGKGTDDIMHB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(S1)C(F)(F)F)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results