accessibility menu, dialog, popup

UserName

H3CBrNCO

CAS RN 207974-15-0

IUPAC Name: 1-bromo-4-(1-isocyanatoethyl)benzene
Synonyms: 1-bromo-4-(1-isocyanatoethyl)benzene
Molecular Weight (g/mol): 226.07
Molecular Formula: C9H8BrNO
InChi Key: AKVAGWOVWQDTAF-UHFFFAOYNA-N
SMILES: CC(N=C=O)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results