accessibility menu, dialog, popup

UserName

CAS RN 207742-76-5

NNNH2H2NH2NOHSOOHO

CAS RN 207742-76-5

IUPAC Name: pyrimidine-4,5,6-triamine sulfuric acid hydrate
Synonyms: pyrimidine-4,5,6-triamine sulfuric acid hydrate
Molecular Weight (g/mol): 241.22
Molecular Formula: C4H11N5O5S
InChi Key: WRXLIMZODSSQIH-UHFFFAOYSA-N
SMILES: O.OS(O)(=O)=O.NC1=NC=NC(N)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results