accessibility menu, dialog, popup

UserName

CAS RN 207511-18-0

H3CONO

CAS RN 207511-18-0

IUPAC Name: 4-methoxypyridin-1-ium-1-olate
Synonyms: 4-methoxypyridin-1-ium-1-olate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: BOFAIBPJCWFJFT-UHFFFAOYSA-N
SMILES: COC1=CC=[N+]([O-])C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results