accessibility menu, dialog, popup

UserName

CAS RN 207398-97-8

IUPAC Name: indium(3+) hydrate trinitrate
Synonyms: indium(3+) hydrate trinitrate
Molecular Weight (g/mol): 318.85
Molecular Formula: H2InN3O10
InChi Key: YZZFBYAKINKKFM-UHFFFAOYSA-N
SMILES: O.[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)