accessibility menu, dialog, popup

UserName

CAS RN 207234-02-4

H3CNCl

CAS RN 207234-02-4

IUPAC Name: 1-dodecylpyridin-1-ium hydrate chloride
Synonyms: 1-dodecylpyridin-1-ium hydrate chloride
Molecular Weight (g/mol): 301.9
Molecular Formula: C17H32ClNO
InChi Key: BDGGUWSWAKGEGH-UHFFFAOYSA-M
SMILES: O.[Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results