CAS RN 2065-70-5
CAS RN 2065-70-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dichloronaphthalene, TRC
CAS: 2065-70-5 Molecular Formula: C10H6Cl2 Molecular Weight (g/mol): 197.06 Synonym: PCN 11 SMILES: Clc1ccc2cc(Cl)ccc2c1