accessibility menu, dialog, popup

UserName

CAS RN 2065-67-0

IUPAC Name: tetraphenylphosphanium iodide
Synonyms: tetraphenylphosphonium iodide
Molecular Weight (g/mol): 466.3
Molecular Formula: C24H20IP
InChi Key: AEFPPQGZJFTXDR-UHFFFAOYSA-M
SMILES: [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results