accessibility menu, dialog, popup

UserName

CAS RN 2062-25-1

HOOFFF(E)

CAS RN 2062-25-1

IUPAC Name: (2E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: (2E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 216.16
Molecular Formula: C10H7F3O2
InChi Key: AMVYAIXPAGBXOM-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results