accessibility menu, dialog, popup

UserName

CAS RN 20595-30-6

HOOF(E)

CAS RN 20595-30-6

IUPAC Name: (2E)-3-(3-fluorophenyl)prop-2-enoic acid
Synonyms: (2E)-3-(3-fluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 166.15
Molecular Formula: C9H7FO2
InChi Key: RTSIUKMGSDOSTI-SNAWJCMRSA-N
SMILES: OC(=O)\C=C\C1=CC=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results