CAS RN 20548-54-3
CAS RN 20548-54-3
IUPAC Name:
sulfanylidenecalcium
Synonyms:
calcium sulfide
Molecular Weight (g/mol):
72.14
Molecular Formula:
CaS
InChi Key:
JGIATAMCQXIDNZ-UHFFFAOYSA-N
SMILES:
S=[Ca]
1
–
2
of
2
results
1
Calcium sulfide, 99.9% (metals basis excluding Sr), Sr typically 500 ppm
CAS: 20548-54-3 Molecular Formula: CaS Molecular Weight (g/mol): 72.14 MDL Number: MFCD00015990 InChI Key: AGVJBLHVMNHENQ-UHFFFAOYSA-N PubChem CID: 30182 IUPAC Name: calcium sulfanediide SMILES: [S--].[Ca++]
2
Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%
CAS: 20548-54-3 Molecular Formula: CaS Molecular Weight (g/mol): 72.14 MDL Number: MFCD00015990 InChI Key: AGVJBLHVMNHENQ-UHFFFAOYSA-N PubChem CID: 30182 IUPAC Name: calcium sulfanediide SMILES: [S--].[Ca++]