CAS RN 205055-71-6
CAS RN 205055-71-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloroquinoline-6-sulfonyl Chloride, TRC
CAS: 205055-71-6 Molecular Formula: C9 H5 Cl2 N O2 S Molecular Weight (g/mol): 262.11 IUPAC Name: 2-chloroquinoline-6-sulfonyl chloride SMILES: Clc1ccc2cc(ccc2n1)S(=O)(=O)Cl