accessibility menu, dialog, popup

UserName

CAS RN 203927-98-4

BCH3CH3BHOOHOHOH

CAS RN 203927-98-4

IUPAC Name: [7-(dihydroxyboranyl)-9,9-dihexyl-9H-fluoren-2-yl]boronic acid
Synonyms: 7-(dihydroxyboranyl)-9,9-dihexylfluoren-2-ylboronic acid
Molecular Weight (g/mol): 422.18
Molecular Formula: C25H36B2O4
InChi Key: QFWOJNOZWMHNTK-UHFFFAOYSA-N
SMILES: CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results