accessibility menu, dialog, popup

UserName

OOClCl

CAS RN 2032-75-9

IUPAC Name: 2,2-dichloro-2H-1,3-benzodioxole
Synonyms: 2,2-dichloro-1,3-benzodioxole
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: NMNKOUVBELKKTR-UHFFFAOYSA-N
SMILES: ClC1(Cl)OC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results