CAS RN 20315-25-7
CAS RN 20315-25-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Procyanidin B1, TRC
CAS: 20315-25-7 Molecular Formula: C30 H26 O12 Molecular Weight (g/mol): 578.52 Synonym: (2R,2'R,3R,3'S,4R)-2,2'-bis(3,4-dihydroxyphenyl)-3,3',4,4'-tetrahydro-[4,8'-Bi-2H-1-benzopyran]-3,3',5,5',7,7'-hexol,(-)-Epicatechin-(4β-8)-(+)-catechin,Proanthocyanidin B1,Procyanidol B1 IUPAC Name: (2R,3R,4R)-2-(3,4-dihydroxyphenyl)-4-[(2R,3S)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-8-yl]-3,4-dihydro-2H-chromene-3,5,7-triol SMILES: O[C@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4cc(O)cc(O)c34)c5ccc(O)c(O)c5)c2O[C@@H]1c6ccc(O)c(O)c6