CAS RN 201990-24-1
CAS RN 201990-24-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
N-(1,1-Dioxido-2,3-dihydro-3-thienyl)glycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(1,1-Dioxido-2,3-dihydro-3-thienyl)glycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(1,1-Dioxido-2,3-dihydro-3-thienyl)glycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.