accessibility menu, dialog, popup

UserName

NH2ClFBr

CAS RN 201849-14-1

IUPAC Name: 2-bromo-6-chloro-4-fluoroaniline
Synonyms: 2-bromo-6-chloro-4-fluoroaniline
Molecular Weight (g/mol): 224.46
Molecular Formula: C6H4BrClFN
InChi Key: LIBGMUMMWYKJSC-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(F)C=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results