accessibility menu, dialog, popup

UserName

CAS RN 20098-14-0

OHO

CAS RN 20098-14-0

IUPAC Name: 5-hydroxyadamantan-2-one
Synonyms: kemantan kamantane kemantane idramantone
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: TZBDEVBNMSLVKT-UHFFFAOYNA-N
SMILES: OC12CC3CC(C1)C(=O)C(C3)C2
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)