accessibility menu, dialog, popup

UserName

CH3CH3H3CSOOHNFFF

CAS RN 200933-14-8

IUPAC Name: 2,4,6-trimethyl-N-[3-(trifluoromethyl)phenyl]benzene-1-sulfonamide
Synonyms: 2,4,6-trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
Molecular Weight (g/mol): 343.36
Molecular Formula: C16H16F3NO2S
InChi Key: ZIIUUSVHCHPIQD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results