CAS RN 1996-30-1
CAS RN 1996-30-1
IUPAC Name:
2-bromo-4-chloro-1-fluorobenzene
Synonyms:
2-bromo-4-chloro-1-fluorobenzene
Molecular Weight (g/mol):
209.44
Molecular Formula:
C6H3BrClF
InChi Key:
YFFUYGSLQXVHMB-UHFFFAOYSA-N
SMILES:
FC1=C(Br)C=C(Cl)C=C1
1
–
1
of
1
results
1
2-Bromo-4-chloro-1-fluorobenzene, 97%
CAS: 1996-30-1 Molecular Formula: C6H3BrClF Molecular Weight (g/mol): 209.442 MDL Number: MFCD00672934 InChI Key: YFFUYGSLQXVHMB-UHFFFAOYSA-N Synonym: 3-bromo-4-fluorochlorobenzene,3-bromo-1-chloro-4-fluorobenzene,2-fluoro-5-chlorobromobenzene,1-bromo-5-chloro-2-fluorobenzene,1-bromo-3-chloro-6-fluorobenzene,5-chloro-2-fluorobromobenzene,2-bromo-4-chloro-1-fluoro-benzene,2-bromo-4-chlorofluorobenzene,benzene, 2-bromo-4-chloro-1-fluoro,pubchem3210 PubChem CID: 2773264 IUPAC Name: 2-bromo-4-chloro-1-fluorobenzene SMILES: C1=CC(=C(C=C1Cl)Br)F