accessibility menu, dialog, popup

UserName

CAS RN 19662-59-0

IUPAC Name: copper(2+) 2-methylprop-2-enoate
Synonyms: copper(2+) methacrylate
Molecular Weight (g/mol): 148.63
Molecular Formula: C4H5CuO2
InChi Key: BFHDHRZKYIRNRP-UHFFFAOYSA-M
SMILES: [Cu++].CC(=C)C([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)