CAS RN 192504-81-7
CAS RN 192504-81-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Desmethyl Nicergoline, TRC
CAS: 192504-81-7 Molecular Formula: C23 H24 Br N3 O3 Molecular Weight (g/mol): 470.36 Synonym: Ergoline-8-methanol, 10-methoxy-1-methyl-, 5-bromo-3-pyridinecarboxylate (ester), (8β)- (9CI),N6-Demethylnicergoline,[(6aR,9R,10aS)-10a-Methoxy-4-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methyl 5-bromopyridine-3-carboxylate,6-Demethylnicergoline,6-Desmethylnicergoline,Nicergoline Imp. H (EP) IUPAC Name: [(6aR,9R,10aS)-10a-methoxy-4-methyl-6,6a,7,8,9,10-hexahydroindolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate SMILES: CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN[C@@H]1Cc4cn(C)c5cccc2c45