accessibility menu, dialog, popup

UserName

H3COONHOONOOCH3

CAS RN 192053-05-7

IUPAC Name: 3-ethyl 5-(3-nitrophenyl)methyl 2-methyl-6-phenyl-4-(2-phenylethynyl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: 3-ethyl 5-(3-nitrophenyl)methyl 2-methyl-6-phenyl-4-(2-phenylethynyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol): 522.56
Molecular Formula: C31H26N2O6
InChi Key: WICWQHSRWZIPNB-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(C)NC(C2=CC=CC=C2)=C(C1C#CC1=CC=CC=C1)C(=O)OCC1=CC(=CC=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results