accessibility menu, dialog, popup

UserName

CAS RN 19099-93-5

IUPAC Name: benzyl 4-oxopiperidine-1-carboxylate
Synonyms: benzyl 4-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 233.27
Molecular Formula: C13H15NO3
InChi Key: VZOVOHRDLOYBJX-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CCC(=O)CC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)