accessibility menu, dialog, popup

UserName

CAS RN 1906-79-2

BrH3CN

CAS RN 1906-79-2

IUPAC Name: 1-ethylpyridin-1-ium bromide
Synonyms: 1-ethylpyridinium bromide
Molecular Weight (g/mol): 188.07
Molecular Formula: C7H10BrN
InChi Key: ABFDKXBSQCTIKH-UHFFFAOYSA-M
SMILES: [Br-].CC[N+]1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results