CAS RN 189340-78-1
CAS RN 189340-78-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
sym-Homo Spermidine Trihydrochloride, TRC
CAS: 189340-78-1 Molecular Formula: C8H21N3•3HCl Molecular Weight (g/mol): 159.2733646 Synonym: N1-(4-Aminobutyl)-1,4-butanediamine Trihydrochloride,4,4'-Diaminodibutylamine Trihydrochloride,4,4'-Iminobis[butylamine] Trihydrochloride,Bis-(4-aminobutyl)amine Trihydrochloride,N-(4-Aminobutyl)-1,4-butanediamine Trihydrochloride,N-(4-Aminobutyl)-1,4-diaminobutane Trihydrochloride,sym-Homospermidine Trihydrochloride SMILES: C(CCNCCCCN)CN.Cl.Cl.Cl