accessibility menu, dialog, popup

UserName

SBrN

CAS RN 18791-98-5

IUPAC Name: 3-bromothiophene-2-carbonitrile
Synonyms: 3-bromothiophene-2-carbonitrile
Molecular Weight (g/mol): 188.04
Molecular Formula: C5H2BrNS
InChi Key: RNIRKEQGYHHNSE-UHFFFAOYSA-N
SMILES: BrC1=C(SC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results