accessibility menu, dialog, popup

UserName

CH3OOONOO

CAS RN 1874-23-3

IUPAC Name: methyl 5-nitrofuran-2-carboxylate
Synonyms: methyl 5-nitrofuran-2-carboxylate
Molecular Weight (g/mol): 171.11
Molecular Formula: C6H5NO5
InChi Key: UTLKCGPAJUYGOM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results