CAS RN 18719-22-7
CAS RN 18719-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-[(4-Chlorophenyl)phenylmethyl]piperazine Hydrochloride, TRC
CAS: 18719-22-7 Molecular Formula: C17H20Cl2N2 • xHCl Molecular Weight (g/mol): 323.26 Synonym: Norchlorcyclizine Hydrochloride IUPAC Name: 1-((4-chlorophenyl)(phenyl)methyl)piperazine hydrochloride SMILES: ClC1=CC=C(C(N2CCNCC2)C3=CC=CC=C3)C=C1.[•HCl]