CAS RN 1858255-62-5
IUPAC Name:
8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Synonyms:
8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Molecular Weight (g/mol):
293.5
Molecular Formula:
C8H6BrClN2O3
InChi Key:
ZROWNTVYDRILKQ-UHFFFAOYSA-N
SMILES:
O.OC(=O)C1=CN2C=C(Cl)C=C(Br)C2=N1
1
–
1
of
1
results
1
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™
CAS: 1858255-62-5 Molecular Formula: C8H6BrClN2O3 Molecular Weight (g/mol): 293.501 MDL Number: MFCD22682816 InChI Key: ZROWNTVYDRILKQ-UHFFFAOYSA-N Synonym: 8-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate PubChem CID: 121235590 IUPAC Name: 8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid;hydrate SMILES: C1=C(C2=NC(=CN2C=C1Cl)C(=O)O)Br.O