accessibility menu, dialog, popup

UserName

CAS RN 185629-31-6

CH3OONOOF

CAS RN 185629-31-6

IUPAC Name: methyl 3-fluoro-4-nitrobenzoate
Synonyms: methyl 3-fluoro-4-nitrobenzoate
Molecular Weight (g/mol): 199.14
Molecular Formula: C8H6FNO4
InChi Key: FKMZNQQOPCCUTD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results