accessibility menu, dialog, popup

UserName

CAS RN 18511-69-8

NNH2NH2N

CAS RN 18511-69-8

IUPAC Name: [2,2'-bipyridine]-4,4'-diamine
Synonyms: [2,2'-bipyridine]-4,4'-diamine
Molecular Weight (g/mol): 186.22
Molecular Formula: C10H10N4
InChi Key: WTHJTVKLMSJXEV-UHFFFAOYSA-N
SMILES: NC1=CC(=NC=C1)C1=NC=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results