accessibility menu, dialog, popup

UserName

CAS RN 18496-54-3

OCl

CAS RN 18496-54-3

IUPAC Name: 4-phenylbutanoyl chloride
Synonyms: 4-phenylbutanoyl chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: VQDQISMDUHBUFF-UHFFFAOYSA-N
SMILES: ClC(=O)CCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results