accessibility menu, dialog, popup

UserName

CAS RN 1840-27-3

CH3FH3CNH2

CAS RN 1840-27-3

IUPAC Name: 4-fluoro-3,5-dimethylaniline
Synonyms: 4-fluoro-3,5-dimethylaniline
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: IGNFILTZSIUWBS-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC(C)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results