accessibility menu, dialog, popup

UserName

Na(S)(R)(R)OOHOHHOOPOOONa

CAS RN 18265-46-8

IUPAC Name: [(2R,3S,4R,5S)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Synonyms: [(2R,3S,4R,5S)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol): 228.09
Molecular Formula: C5H9O8P
InChi Key: KTVPXOYAKDPRHY-AIHAYLRMSA-L
SMILES: O[C@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results