accessibility menu, dialog, popup

UserName

CH3SiH3CCH3S

CAS RN 18245-28-8

IUPAC Name: trimethyl(thiophen-2-yl)silane
Synonyms: trimethyl(thiophen-2-yl)silane
Molecular Weight (g/mol): 156.32
Molecular Formula: C7H12SSi
InChi Key: OANGLGSZPSFVDY-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results