accessibility menu, dialog, popup

UserName

HOBOHOO

CAS RN 182281-01-2

IUPAC Name: [4-(oxan-2-yloxy)phenyl]boronic acid
Synonyms: 4-(oxan-2-yloxy)phenylboronic acid
Molecular Weight (g/mol): 222.05
Molecular Formula: C11H15BO4
InChi Key: DMGMCZRNMYQQQB-UHFFFAOYNA-N
SMILES: OB(O)C1=CC=C(OC2CCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results